missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TRPM7 Antibody (S74), DyLight 488, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-12906G
This item is not returnable.
View return policy
Description
TRPM7 Monoclonal specifically detects TRPM7 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications
| TRPM7 | |
| Monoclonal | |
| DyLight 488 | |
| CHAK, CHAK1FLJ25718, Channel-kinase 1, EC 2.7.11.1, Long transient receptor potential channel 7, LTRPC ion channel family member 7, LTrpC7, LTrpC-7, LTRPC7FLJ20117, transient receptor potential cation channel subfamily M member 7, transient receptor potential cation channel, subfamily M, member 7, transient receptor potential-phospholipase C-interacting kinase, TRP-PLIK | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| Detects approx 220kDa. No cross-reactivity against TrpM6. | |
| Store at 4C in the dark. | |
| IgG1 |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| S74-25 | |
| Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin | |
| TRPM7 | |
| Fusion protein amino acids 1817-1863 (C- terminus) of mouse TrpM7 | |
| 0.1 mL | |
| Neurodegeneration, Neuronal Cell Markers, Neuroscience, Neurotransmission | |
| 54822 | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction