missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TRPM8 Antibody, CoraFluor™ 1, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB200-145CL1
This item is not returnable.
View return policy
Description
TRPM8 Polyclonal antibody specifically detects TRPM8 in Drosophila,Human,Mouse samples. It is validated for ELISA,Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin),Immunohistochemistry (Frozen),Immunocytochemistry/Immunofluorescence
Specifications
| TRPM8 | |
| Polyclonal | |
| PBS | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Drosophila, Human, Mouse | |
| Purified |
| ELISA, Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen) | |
| CoraFluor 1 | |
| Long transient receptor potential channel 6, LTrpC-6, LTRPC6, MGC2849, short form of the TRPM8 cationic channel, transient receptor potential cation channel, subfamily M, member 8, Transient receptor potential p8, transient receptor potential subfamily M member 8, trp-p8, TRPP8transient receptor potential cation channel subfamily M member 8 | |
| Synthetic peptides made to residues 278-292 RNQLEKYISERTIQD and the C-terminus sequence NDLKGLLKEIANKIK of the human trp-p8 protein. | |
| 0.1 mL | |
| Cancer, Tumor Suppressors | |
| 79054 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction