missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TRPM8 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5143781
This item is not returnable.
View return policy
Description
This antibody detects a 130 kDa* protein on SDS-PAGE immunoblots of human MDA-MB-231 and MCF-7 cells. The antibody is also recommended for immunocytochemical labeling of cells.
TRPM8 is a receptor-activated non-selective cation channel involved in detection of sensations such as coolness, by being activated by cold temperature below 25 degrees Celsius. TRPM8 is activated by icilin, eucalyptol, menthol, cold and modulation of intracellular pH and involved in menthol sensation. TRPM8 is permeable for monovalent cations sodium, potassium, and cesium and divalent cation calcium. TRPM8 has an expression pattern within normal human tissues that is mostly restricted to prostate epithelial cells. TRPM8 mRNA expression is at moderate levels in normal prostate tissue and appears to be elevated in prostate cancer. Notably, TRPM8 mRNA was also expressed in a number of nonprostatic primary tumors of breast, colon, lung, and skin origin, whereas transcripts encoding TRPM8 were hardly detected or not detected in the corresponding normal human tissues.
Specifications
| TRPM8 | |
| Polyclonal | |
| Unconjugated | |
| TRPM8 | |
| CMR1; Cold menthol receptor 1; cold/menthol receptor 1; long transient receptor potential channel 6; LTRPC6; LTrpC-6; MGC2849; short form of the TRPM8 cationic channel; transient receptor melastatin 8 variant 1; transient receptor melastatin 8 variant 2; transient receptor potential cation channel subfamily M member 8; transient receptor potential cation channel, subfamily M, member 8; transient receptor potential p8; transient receptor potential subfamily M member 8; transient receptor potential-p8; TRP ion channel TRPM8; Trpm8; Trpp8; trp-p8 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 171382, 171384, 79054 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 1mg/mL BSA, 50% glycerol and 0.05% sodium azide | |
| Q7Z2W7, Q8R455, Q8R4D5 | |
| TRPM8 | |
| TRPM8 synthetic peptide (coupled to carrier) corresponding to amino acids in the extracellular region of human TRPM8. This site is well conserved in rat and mouse TRPM8, but has low homology to other TRPM family members. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction