missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TRPS1 Antibody (TRPS1/7912R), DyLight 650, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-21051C
This item is not returnable.
View return policy
Description
TRPS1 Monoclonal antibody specifically detects TRPS1 in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| TRPS1 | |
| Monoclonal | |
| DyLight 650 | |
| trichorhinophalangeal syndrome I, tricho-rhino-phalangeal syndrome type I protein | |
| Recombinant fragment (around aa900-1100) of human TRPS1 protein (exact sequence is proprietary) | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Immunohistochemistry (Paraffin) | |
| TRPS1/7912R | |
| 50 mM Sodium Borate | |
| Rabbit | |
| Protein A or G purified | |
| RUO | |
| 7227 | |
| Store at 4°C in the dark. | |
| IgG κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction