missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TRPV1 Polyclonal Antibody, OriGene
Rabbit Polyclonal Antibody
Brand: Origene Technologies TA890016S
This item is not returnable.
View return policy
Description
TRPV1 Polyclonal antibody specifically detects TRPV1 in Human samples. It is validated for Western BlotSpecifications
| TRPV1 | |
| Polyclonal | |
| Unconjugated | |
| FANCL | |
| Capsaicin receptor; cation channel; DKFZp434K0220; non-selective cation channel; osm-9-like TRP channel 1; OTRPC1; si:dkey-9e15.2; transient receptor potential cation channel subfamily V member 1; transient receptor potential cation channel V1; transient receptor potential cation channel, subfamily V, member 1; transient receptor potential V1; transient receptor potential V1 channel; transient receptor potential vanilloid 1 supraoptic nucleus; transient receptor potential vanilloid 1a; transient receptor potential vanilloid 1b; TRPV1; TRPV1/2; TRPV1_SON; TRPV1alpha; TRPV1beta; Vanilloid receptor; vanilloid receptor 1; vanilloid receptor subtype 1; vanilloid receptor type 1 like protein 1; Vanilloid receptor type 1-like; vanilloid type 1 receptor; VR.5'sv; Vr1; VR-1; Vr1l; Vr1l1 | |
| Recombinant protein of human FANCL | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
| Western Blot | |
| 1.48 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide | |
| Q9NW38 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 55120 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction