missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TRPV4 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
€ 189.00 - € 550.00
Specifications
| Antigen | TRPV4 |
|---|---|
| Concentration | 1 mg/mL |
| Applications | Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18674379
|
Novus Biologicals
NBP2-41262-0.025mg |
0.025 mg |
€ 189.00
0.03mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18081386
|
Novus Biologicals
NBP2-41262 |
0.1 mg |
€ 550.00
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
TRPV4 Polyclonal specifically detects TRPV4 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
| TRPV4 | |
| Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| RUO | |
| PBS with 0.02% Sodium Azide | |
| 59341 | |
| Antibody was raised against an 18 amino acid peptide near the center of human TRPV4. The immunogen is located within amino acids 380 - 430 of TRPV4. Amino Acid Squence: RREVTDEDTRHLSRKFKD | |
| Primary | |
| TRPV4 antibody is human specific. At least six isoforms of TRPV4 are known to exist. This antibody is predicted to not cross-react with other TRP protein family members. |
| 1 mg/mL | |
| Polyclonal | |
| Rabbit | |
| Human | |
| CMT2Cosmosensitive transient receptor potential channel 4, HMSN2C, Osm-9-like TRP channel 4, OTRPC4SSQTL1, transient receptor potential cation channel subfamily V member 4, transient receptor potential cation channel, subfamily V, member 4, Transient receptor potential protein 12, TRP12OSM9-like transient receptor potential channel 4, TrpV4, Vanilloid receptor-like channel 2, Vanilloid receptor-like protein 2, vanilloid receptor-related osmotically activated channel, Vanilloid receptor-related osmotically-activated channel, VRL2, VRL-2, VR-OAC, VROACSPSMA | |
| TRPV4 | |
| IgG | |
| Affinity Purified | |
| 100 kDa |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title