missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ TRPV6 Overexpression Lysate

Product Code. 18265980 Shop All Bio Techne Products
Click to view available options
:
0.1mg
This item is not returnable. View return policy

Product Code. 18265980

Brand: Novus Biologicals™ NBL117345

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
The TRPV6 Overexpression Lysate is derived from human. It has been validated for the following applications: Western Blot.
TRUSTED_SUSTAINABILITY

Specifications

Cell Line The lysate was created in HEK293T cells, using plasmid ID RC214982 and based on accession number NM_18646. The protein contains a C-terminal DDK tag.
For Use With (Application) Western Blot
Quantity 0.1 mg
Storage Buffer RIPA buffer
Purity Protein
Test Specificity Homo sapiens transient receptor potential cation channel, subfamily V, member 6 (TRPV6), mRNA.
Content And Storage -80°C. Avoid freeze-thaw cycles.
Protein State Native

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.