Learn More
TRRAP Antibody, DyLight 488, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand:  Novus Biologicals NB100-58810G
Code produit. 18769311
  • € 633.00 / 0.10mL
Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.
Description
Description
TRRAP Polyclonal antibody specifically detects TRRAP in Human samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY
Spécification
Spécification
TRRAP
Polyclonal
Western Blot
350/400 kDa PCAF-associated factor, FLJ10671, PAF350/400, PAF400STAF40, Tra1, Tra1 homolog, transformation/transcription domain-associated protein, TR-AP
The immunogen recognized by this antibody maps to a region between residue 3275 and 3325 of human transformation/transcription domain-associated protein using the numbering given in entry AAD04629.1 (GeneID 8295).
0.1 mL
Cell Cycle and Replication, Protein Kinase
8295
Store at 4C in the dark.
IgG
Western Blot
DyLight 488
50mM Sodium Borate
Rabbit
Immunogen affinity purified