missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Trypsin Inhibitor Antibody, Alexa Fluor™ 405, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-78088AF405
This item is not returnable.
View return policy
Description
Trypsin Inhibitor Polyclonal antibody specifically detects Trypsin Inhibitor in Plant samples. It is validated for Western Blot, ELISA
Specifications
| Trypsin Inhibitor | |
| Polyclonal | |
| Western Blot, ELISA | |
| KTI3, Kunitz trypsin inhibitor | |
| Trypsin Inhibitor [Lima Bean] (Uniprot: P01070) | |
| 0.1 mL | |
| Primary | |
| Plant | |
| IgG |
| Western Blot, ELISA | |
| Alexa Fluor 405 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Multi-step | |
| RUO | |
| 547831 | |
| Store at 4C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction