missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tryptase alpha/beta 1 Antibody (RM0355-10H34), Alexa Fluor™ 488, Novus Biologicals™
Rat Monoclonal Antibody
Brand: Novus Biologicals NBP2-11983AF488
This item is not returnable.
View return policy
Description
Tryptase alpha/beta 1 Monoclonal antibody specifically detects Tryptase alpha/beta 1 in Mouse samples. It is validated for Western Blot.
Specifications
| Tryptase alpha/beta 1 | |
| Monoclonal | |
| Alexa Fluor 488 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| EC:3.4.21.59, epididymis secretory sperm binding protein, mast cell alpha II tryptase, mast cell beta I tryptase, mast cell tryptase, TPS1, TPS2, TPSB1, TPSB2, tryptase alpha II, Tryptase alpha-1, tryptase beta I, tryptase beta-1, Tryptase beta-2, Tryptase I, tryptase II, tryptase-1, tryptase-2, tryptase-I, tryptase-III | |
| Rat | |
| Protein A or G purified | |
| Immunology, metabolism | |
| 7177 | |
| Store at 4C in the dark. | |
| IgG2 |
| Western Blot | |
| RM0355-10H34 | |
| Western Blot | |
| Q02844 | |
| TPSAB1 | |
| Mouse recombinant Tryptase alpha/beta 1 | |
| 0.1 mL | |
| Primary | |
| Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction