missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tryptase beta-2/TPSB2 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-05583-100ul
This item is not returnable.
View return policy
Description
Tryptase beta-2/TPSB2 Polyclonal antibody specifically detects Tryptase beta-2/TPSB2 in Human, Rat samples. It is validated for Western Blot
Specifications
| Tryptase beta-2/TPSB2 | |
| Polyclonal | |
| Western Blot 1:500-1:2000 | |
| EC 3.4.21, EC 3.4.21.59, mast cell tryptase beta II, mast cell tryptase beta III, TPS2tryptase beta-2, tryptase beta 2, tryptase beta 2 (gene/pseudogene), tryptase beta II, tryptase beta III, Tryptase II, tryptase III, tryptase-2, tryptaseB, tryptaseC | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-275 of human Tryptase beta-2/TPSB2 (NP_077078.5). MLNLLLLALPVLASRAYAAPAPGQALQRVGIVGGQEAPRSKWPWQVSLRVRDRYWMHFCGGSLIHPQWVLTAAHCVGPDVKDLAALRVQLREQHLYYQDQLLPVSRIIVHPQFYTAQIGADIALLELEEPVNVSSHVHTVTLPPASETFPPGMPCWVTGWGDVDNDERLPPPFPLKQVKVPIMENHICDAKYHLGAYTGDDVRIVRDDMLCAGNTRRDSCQGDSGGPLVCKVNGTWLQAGVVSWGEGCAQPNRPGIYTRVTYYLDWIHHYVPKKP | |
| 100 μg | |
| Cell Biology, Immunology, Innate Immunity | |
| 64499 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS with 50% glycerol, pH7.3. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction