missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tryptase gamma-1/TPSG1 Antibody (OTI1E11), FITC, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-74649F
This item is not returnable.
View return policy
Description
Tryptase gamma-1/TPSG1 Monoclonal specifically detects Tryptase gamma-1/TPSG1 in Human samples. It is validated for Flow Cytometry.
Specifications
| Tryptase gamma-1/TPSG1 | |
| Monoclonal | |
| FITC | |
| PBS with 0.05% Sodium Azide | |
| TPSG1 | |
| Recombinant protein fragment corresponding to amino acids 20-283 of human Tryptase gamma-1/TPSG1 (NP_036599) produced in E.coli. | |
| 0.1 mL | |
| 25823.0 | |
| Store at 4°C in the dark. |
| Flow Cytometry | |
| OTI1E11 | |
| Flow Cytometry | |
| EC 3.4.21, EC 3.4.21.-, gamma I, gamma II, lung tryptase, mast cell protease II, mast cell tryptase, PRSS31skin tryptase, Serine protease 31, TMTpituitary tryptase, Transmembrane tryptase, trpA, tryptase gamma, tryptase gamma 1, tryptase gamma I, tryptase gamma II | |
| Mouse | |
| Affinity Purified | |
| Primary | |
| Human | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction