Learn More
Tryptophan EP Impurity A, Mikromol™
Product Code. 30050164
Change view
Click to view available options
Quantity:
25 mg
100 mg
Unit Size:
100mg
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30050164 25 mg 25mg
30052537 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 30050164 Supplier Mikromol Supplier No. MM0172.090025
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
Tryptophan EP Impurity A
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 1,1'-Ethylidenebistryptophan
CAS 132685-02-0
Analyte or Component Names 1,1'-Ethylidenebistryptophan
Shipping Condition Ice pack (-20°C)
Molecular Formula C24 H26 N4 O4
Certifications/Compliance ISO 9001
Synonym 3,3'-[Ethylidenebis(1H-indole-1,3-diyl)]bis[(2S)-2-aminopropanoic] Acid (1,1'-Ethylidenebistryptophan), Tryptophan Imp. A (EP), Tryptophan USP Related Compound A, Tryptophan USP RC A, 3,3'-[Ethylidenebis(1H-indole-1,3-diyl)]bis[(2S)-2-aminopropanoic] Acid, 1,1'-Ethylidenebistryptophan, Tryptophan Related Compound A, Tryptophan Impurity A
InChI Formula InChI=1S/C24H26N4O4/c1-14(27-12-15(10-19(25)23(29)30)17-6-2-4-8-21(17)27)28-13-16(11-20(26)24(31)32)18-7-3-5-9-22(18)28/h2-9,12-14,19-20H,10-11,25-26H2,1H3,(H,29,30)(H,31,32)/t19-,20-/m0/s1
SMILES CC(n1cc(C[C@H](N)C(=O)O)c2ccccc12)n3cc(C[C@H](N)C(=O)O)c4ccccc34
IUPAC Name (2S)-2-amino-3-[1-[1-[3-[(2S)-2-amino-2-carboxyethyl]indol-1-yl]ethyl]indol-3-yl]propanoic acid
Molecular Weight (g/mol) 434.49
Quantity 25 mg
Formula Weight 434.1954 g/mol