Learn More
Tryptophan EP Impurity A, Mikromol™
Description
Tryptophan EP Impurity A
Specifications
Specifications
| Chemical Name or Material | 1,1'-Ethylidenebistryptophan |
| CAS | 132685-02-0 |
| Analyte or Component Names | 1,1'-Ethylidenebistryptophan |
| Shipping Condition | Ice pack (-20°C) |
| Molecular Formula | C24 H26 N4 O4 |
| Certifications/Compliance | ISO 9001 |
| Synonym | 3,3'-[Ethylidenebis(1H-indole-1,3-diyl)]bis[(2S)-2-aminopropanoic] Acid (1,1'-Ethylidenebistryptophan), Tryptophan Imp. A (EP), Tryptophan USP Related Compound A, Tryptophan USP RC A, 3,3'-[Ethylidenebis(1H-indole-1,3-diyl)]bis[(2S)-2-aminopropanoic] Acid, 1,1'-Ethylidenebistryptophan, Tryptophan Related Compound A, Tryptophan Impurity A |
| InChI Formula | InChI=1S/C24H26N4O4/c1-14(27-12-15(10-19(25)23(29)30)17-6-2-4-8-21(17)27)28-13-16(11-20(26)24(31)32)18-7-3-5-9-22(18)28/h2-9,12-14,19-20H,10-11,25-26H2,1H3,(H,29,30)(H,31,32)/t19-,20-/m0/s1 |
| SMILES | CC(n1cc(C[C@H](N)C(=O)O)c2ccccc12)n3cc(C[C@H](N)C(=O)O)c4ccccc34 |
| IUPAC Name | (2S)-2-amino-3-[1-[1-[3-[(2S)-2-amino-2-carboxyethyl]indol-1-yl]ethyl]indol-3-yl]propanoic acid |
| Show More |