missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tryptophan Hydroxylase 1/TPH-1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
€ 271.00 - € 572.00
Specifications
| Antigen | Tryptophan Hydroxylase 1/TPH-1 |
|---|---|
| Dilution | Western Blot -Validated from a verified customer review, Immunohistochemistry 1:50 - 1:200, Immunohistochemistry-Paraffin 1:50 - 1:200 |
| Applications | Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18430881
|
Novus Biologicals
NBP1-86922-25ul |
25 μL |
€ 271.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18421671
|
Novus Biologicals
NBP1-86922 |
0.1 mL |
€ 572.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Tryptophan Hydroxylase 1/TPH-1 Polyclonal antibody specifically detects Tryptophan Hydroxylase 1/TPH-1 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
| Tryptophan Hydroxylase 1/TPH-1 | |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Lipid and Metabolism, Neuroscience, Neurotransmission | |
| PBS (pH 7.2) and 40% Glycerol | |
| 7166 | |
| IgG | |
| Immunogen affinity purified |
| Western Blot -Validated from a verified customer review, Immunohistochemistry 1:50 - 1:200, Immunohistochemistry-Paraffin 1:50 - 1:200 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human | |
| EC 1.14.16, EC 1.14.16.4, indoleacetic acid-5-hydroxylase, L-tryptophan hydroxylase, MGC119994, TPH, TPRH, TRPH, tryptophan 5-hydroxylase 1, Tryptophan 5-monooxygenase 1, tryptophan hydroxylase (tryptophan 5-monooxygenase), tryptophan hydroxylase 1 | |
| This antibody was developed against Recombinant Protein corresponding to amino acids: ISELKHALSGHAKVKPFDPKITCKQECLITTFQDVYFVSESFEDAKEKMREFTKTIKRPFGVKYNPYTRSIQILKDTKSITSAMNELQHDLDVVSDALAKVSRKPSI | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title