missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tryptophan Hydroxylase 1/TPH-1 Antibody, Novus Biologicals™
Sheep Polyclonal Antibody has been used in 2 publications
Brand: Novus Biologicals NB300-176
This item is not returnable.
View return policy
Description
Tryptophan Hydroxylase 1/TPH-1 Polyclonal specifically detects Tryptophan Hydroxylase 1/TPH-1 in Human, Mouse, Rat, Rabbit (Negative) samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Frozen.
Specifications
| Tryptophan Hydroxylase 1/TPH-1 | |
| Polyclonal | |
| Western Blot 1:1000, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Frozen 1:1000 | |
| P17752 | |
| TPH1 | |
| Recombinant rabbit tryptophan hydroxylase, isolated as inclusion bodies from E. coli and purified by preparative SDS-PAGE. Accession # P17290 | |
| Antigen Affinity-purified | |
| RUO | |
| Primary | |
| Specific for the ∼55k tryptophan hydroxylase protein. | |
| Store at -20C. Avoid freeze-thaw cycles. |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Frozen) | |
| Unconjugated | |
| 10mM HEPES (pH 7.5), 0.15M NaCl, 0.1 mg/ml BSA and 50% Glycerol with No Preservative | |
| EC 1.14.16, EC 1.14.16.4, indoleacetic acid-5-hydroxylase, L-tryptophan hydroxylase, MGC119994, TPH, TPRH, TRPH, tryptophan 5-hydroxylase 1, Tryptophan 5-monooxygenase 1, tryptophan hydroxylase (tryptophan 5-monooxygenase), tryptophan hydroxylase 1 | |
| Sheep | |
| 55 kDa | |
| 0.1 mL | |
| Lipid and Metabolism, Neuroscience, Neurotransmission | |
| 7166 | |
| Human, Mouse, Rat, Rabbit (Negative) | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction