missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tryptophan hydroxylase 2 Antibody, Janelia Fluor™ 549, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-77559JF549
This item is not returnable.
View return policy
Description
Tryptophan hydroxylase 2 Polyclonal antibody specifically detects Tryptophan hydroxylase 2 in Human samples. It is validated for Western Blot
Specifications
| Tryptophan hydroxylase 2 | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot | |
| Janelia Fluor 549 | |
| ADHD7, EC 1.14.16, EC 1.14.16.4, FLJ37295, MGC138872, Neuronal tryptophan hydroxylase, NTPHMGC138871, tryptophan 5-hydroxylase 2, Tryptophan 5-monooxygenase 2, tryptophan hydroxylase 2 | |
| A synthetic peptide made to an N-terminal portion of the human Tryptophan hydroxylase 2 protein (between residues 100-200) [UniProt# Q8IWU9] | |
| 0.1 mL | |
| Lipid and Metabolism | |
| 121278 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction