missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tryptophan hydroxylase 2 Antibody, PerCP, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB100-74555PCP
This item is not returnable.
View return policy
Description
Tryptophan hydroxylase 2 Polyclonal antibody specifically detects Tryptophan hydroxylase 2 in Human, Mouse, Rat, Primate, Rabbit samples. It is validated for Flow Cytometry
Specifications
| Tryptophan hydroxylase 2 | |
| Polyclonal | |
| PBS | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat, Primate, Rabbit | |
| Purified |
| Flow Cytometry | |
| PerCP | |
| ADHD7, EC 1.14.16, EC 1.14.16.4, FLJ37295, MGC138872, Neuronal tryptophan hydroxylase, NTPHMGC138871, tryptophan 5-hydroxylase 2, Tryptophan 5-monooxygenase 2, tryptophan hydroxylase 2 | |
| Synthetic peptide make to an internal portion of the mouse Tryptophan hydroxylase 2 protein (between amino acids 10-75) [UniProt Q8CGV2] | |
| 0.1 mL | |
| Lipid and Metabolism | |
| 121278 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction