missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tryptophan rich protein Antibody, mFluor Violet 450 SE, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-97813MFV450
This item is not returnable.
View return policy
Description
Tryptophan rich protein Polyclonal antibody specifically detects Tryptophan rich protein in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| Tryptophan rich protein | |
| Polyclonal | |
| Immunohistochemistry-Paraffin | |
| congenital heart disease 5 protein10tryptophan-rich protein, FLJ51808, tryptophan rich basic protein | |
| Produced in rabbits immunized with E. coli-derived Human Tryptophan rich protein fragment. | |
| 0.1 mL | |
| Neurodegeneration, Neuroscience | |
| 7485 | |
| Store at 4C in the dark. | |
| IgG |
| Immunohistochemistry (Paraffin) | |
| mFluor Violet 450 SE | |
| 50mM Sodium Borate | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction