missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tryptophanyl tRNA synthetase Antibody, DyLight 488, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-32186G
This item is not returnable.
View return policy
Description
Tryptophanyl tRNA synthetase Polyclonal antibody specifically detects Tryptophanyl tRNA synthetase in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
| Tryptophanyl tRNA synthetase | |
| Polyclonal | |
| Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin | |
| EC 6.1.1, EC 6.1.1.2, GAMMA-2, hWRS, IFI53, IFP53trpRS, Interferon-induced protein 53, TrpRS, tryptophan tRNA ligase 1, cytoplasmic, Tryptophan--tRNA ligase, tryptophanyl-tRNA synthetase, tryptophanyl-tRNA synthetase, cytoplasmic, WRS | |
| The immunogen this antibody was made to, maps to a region between residue 421 to 471 of human Tryptophanyl-tRNA Synthetase using the numbering given in entry NP_004175.2 (GeneID 7453). | |
| 0.1 mL | |
| Cell Cycle and Replication | |
| 7453 | |
| Store at 4°C in the dark. | |
| IgG |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| DyLight 488 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction