missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TSC1 Antibody (488915) [Janelia Fluor™ 585], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB4379JF585
This item is not returnable.
View return policy
Description
TSC1 Monoclonal antibody specifically detects TSC1 in Human samples. It is validated for Western Blot,Flow Cytometry (Intracellular Staining),CyTOF
Specifications
| TSC1 | |
| Monoclonal | |
| Janelia Fluor 585 | |
| KIAA0243MGC86987, LAMhamartin, TSC, tuberous sclerosis 1, Tuberous sclerosis 1 protein, tumor suppressor | |
| E. coli-derived recombinant human TSC1, Asp156-Thr300, Accession # Q92574 | |
| RUO | |
| Primary | |
| Human | |
| Liquid |
| Western Blot, Flow Cytometry (Intracellular Staining), CyTOF | |
| 488915 | |
| 50mM Sodium Borate | |
| Mouse | |
| 0.1 mL | |
| Autophagy, Cancer, Hypoxia, mTOR Pathway | |
| 7248 | |
| Store at 4°C in the dark. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction