missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TSC1 Antibody, Alexa Fluor™ 350, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-76618AF350
This item is not returnable.
View return policy
Description
TSC1 Polyclonal antibody specifically detects TSC1 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunofluorescence
Specifications
| TSC1 | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Rabbit | |
| Peptide affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot, ELISA, Immunofluorescence | |
| Alexa Fluor 350 | |
| KIAA0243MGC86987, LAMhamartin, TSC, tuberous sclerosis 1, Tuberous sclerosis 1 protein, tumor suppressor | |
| Antibody was raised against a 15 amino acid synthetic peptide from the middle region of human TSC1. The immunogen is located within amino acids 220 - 270 of TSC1. Amino Acid Squence: KDHELDPRRWKRLE | |
| 0.1 mL | |
| Autophagy, Cancer, Hypoxia, mTOR Pathway | |
| 7248 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction