missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TSC1 Monoclonal Antibody (OTI2B2), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA802012
This item is not returnable.
View return policy
Description
Implicated as a tumor suppressor. May have a function in vesicular transport. Interaction between TSC1 and TSC2 may facilitate vesicular docking.Defects in TSC1 are the cause of tuberous sclerosis complex (TSC). The molecular basis of TSC is a functional impairement of the hamartin-tuberin complex. TSC is an autosomal domit multi-system disorder that affects especially the brain, kidneys, heart, and skin. TSC is characterized by hamartomas (benign overgrowths predomitly of a cell or tissue type that occurs normally in the organ) and hamartias (developmental abnormalities of tissue combination). Clinical symptoms can range from benign hypopigmented macules of the skin to profound mental retardation with intractable seizures to premature death from a variety of disease-associated causes.Defects in TSC1 may be a cause of focal cortical dysplasia of Taylor balloon cell type (FCDBC). FCDBC is a subtype of cortical displasias linked to chronic intractable epilepsy. Cortical dysplasias display a broad spectrum of structural changes, which appear to result from changes in proliferation, migration, differentiation, and apoptosis of neuronal precursors and neurons during cortical development.Specifications
| TSC1 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q92574 | |
| TSC1 | |
| Human recombit protein fragment corresponding to amino acids 836-1164 of human TSC1 produced in E.coli. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG2a |
| Western Blot | |
| OTI2B2 | |
| Unconjugated | |
| TSC1 | |
| LAM, TSC | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 7248 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction