missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TSC2 Antibody, mFluor Violet 500 SE, Novus Biologicals™
Rabbit Polyclonal Antibody
€ 622.00
Specifications
| Antigen | TSC2 |
|---|---|
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | mFluor Violet 500 SE |
| Form | Purified |
Description
TSC2 Polyclonal antibody specifically detects TSC2 in Human, Mouse samples. It is validated for Western Blot, ELISA, Immunocytochemistry/ ImmunofluorescenceSpecifications
| TSC2 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human | |
| LAMFLJ43106, TSC4, tuberin, tuberous sclerosis 2, Tuberous sclerosis 2 protein | |
| Antibody was raised against a 14 amino acid synthetic peptide from near the amino terminus of human TSC2. The immunogen is located within the first 50 amino acids of TSC2. Amino Acid Squence: TPRPNPRSAEGKQT | |
| Primary | |
| Store at 4°C in the dark. |
| Western Blot | |
| mFluor Violet 500 SE | |
| Rabbit | |
| Apoptosis, Autophagy, Cancer, Cell Biology, Cell Cycle and Replication, Cellular Markers, Hypoxia, Inhibitors Activators and Regulators, mTOR Pathway, Phospho Specific, Signal Transduction, Tumor Suppressors | |
| 50mM Sodium Borate | |
| 7249 | |
| IgG | |
| Peptide affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title