missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TSC22/TSC22D1 Antibody (OTI4H8), Alexa Fluor™ 750, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-74654AF750
This item is not returnable.
View return policy
Description
TSC22/TSC22D1 Monoclonal specifically detects TSC22/TSC22D1 in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry.
Specifications
| TSC22/TSC22D1 | |
| Monoclonal | |
| Alexa Fluor 750 | |
| 50 mM sodium borate with 0.05% sodium azide | |
| TSC22D1 | |
| Full length recombinant protein of human TSC22/TSC22D1 (NP_006013) produced in HEK293T cells. | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Rat | |
| IgG2a |
| Western Blot, Flow Cytometry, Immunohistochemistry | |
| OTI4H8 | |
| Western Blot, Flow Cytometry, Immunohistochemistry | |
| Cerebral protein 2, KIAA1994, MGC17597, Regulatory protein TSC-22, RP11-269C23.2, TGFB1I4, TGFB-stimulated clone 22 homolog, transforming growth factor beta 1 induced transcript 4, Transforming growth factor beta-1-induced transcript 4 protein, transforming growth factor beta-stimulated protein TSC-22, TSC22 domain family 1, TSC22 domain family protein 1, TSC22 domain family, member 1, TSC22DKFZp686O19206 | |
| Mouse | |
| Affinity Purified | |
| Cancer | |
| 8848.0 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction