missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TSC22D2 Antibody, DyLight 755, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-98553IR
This item is not returnable.
View return policy
Description
TSC22D2 Polyclonal antibody specifically detects TSC22D2 in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| TSC22D2 | |
| Polyclonal | |
| 50 mM Sodium Borate | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Immunohistochemistry (Paraffin) | |
| DyLight 755 | |
| KIAA0669TILZ4a, TILZ4, TILZ4b, TILZ4c, TSC22 domain family 2, TSC22 domain family protein 2, TSC22 domain family, member 2, TSC22-related-inducible leucine zipper protein 4 | |
| Produced in rabbits immunized with E. coli-derived Human TSC22D2 fragment. | |
| 0.1 mL | |
| Cancer | |
| 9819 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction