missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ TSC22D2 Polyclonal Antibody
GREENER_CHOICE

Product Code. 15817063
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15817063 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15817063 Supplier Invitrogen™ Supplier No. PA568648

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

This target displays homology in the following species: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Zebrafish: 92%.

TSC22D2 belongs to the TSC-22/Dip/Bun family and the function remains unknown.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TSC22D2
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene TSC22D2
Gene Accession No. 0
Gene Alias 1810043J12Rik; 5530402M19Rik; cb869; id:ibd5097; kiaa0669; KIAA0669-like protein; TILZ4; TILZ4a; TILZ4b; TILZ4c; TSC22 domain family 2; TSC22 domain family member 2; TSC22 domain family protein 2; TSC22 domain family protein 2-like protein; TSC22 domain family, member 2; TSC22D2; tsc22d2 protein; TSC22-related-inducible leucine zipper protein 4; wu:fa55e04; zgc:92475
Gene Symbols TSC22D2
Host Species Rabbit
Immunogen synthetic peptide directed towards the middle region of human TSC22D2.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 499624
Target Species Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.