Learn More
TSC22D3 Mouse anti-Human, Clone: OTI1E11, lyophilized, TrueMAB™
Mouse Monoclonal Antibody
Brand: Origene Technologies CF505368
Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
The TSC22 domain family member 3 protein (TSC22D3) is a leucine zipper protein that functions as a transcriptional regulator. The expression of TSC22D3 is stimulated by glucocorticoids and IL-10 and is thought to play a key role in the anti-inflammatory and immunosuppressive effects of these molecules. TSC22D3 can physically interact with and inhibit the activities of key inflammatory signaling mediators such NF-Kappa-B and AP-1. TSC22D3 functions as a transcriptional co-activator for various nuclear receptors and NF-Kappa-B. It has also been shown to be involved in the differentiation of mesenchymal stem cells towards osteoblasts and bone formation.Specifications
| TSC22D3 | |
| Monoclonal | |
| Unconjugated | |
| TSC22D3 | |
| DIP, DSIPI, GILZ, TSC-22R | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 1831 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Lyophilized |
| Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
| OTI1E11 | |
| PBS with 8% trehalose and no preservative; pH 7.3 | |
| Q99576 | |
| TSC22D3 | |
| Full length human recombit protein of human TSC22D3 produced in HEK293T cell. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.