missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ TSC22D4 Polyclonal Antibody
GREENER_CHOICE

Product Code. 15770291 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15770291 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15770291 Supplier Invitrogen™ Supplier No. PA552342

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Immunogen sequence: TPLPSLRVEA EAGGSGARTP PLSRRKAVDM RLRMELGAPE EMGQVPPLDS RPSSPALYFT HDASLVHKSP DPFGAVAAQK FSLAHSMLAI SGHLDSDDDS GSGSLVGIDN KIEQAMDLVK SHLMF Highest antigen sequence identity to the following orthologs: Mouse - 82%, Rat - 82%.

TSC22D4 is a member of the TSC22 domain family of leucine zipper transcriptional regulators. Gene Ontology (GO) annotations related to this gene include DNA-binding transcription factor activity. An important paralog of this gene is TSC22D1. Binds DNA and acts as a transcriptional repressor (PubMed:10488076). Involved in the regulation of systematic glucose homeostasis and insulin sensitivity, via transcriptional repression of downstream insulin signaling targets such as OBP2A/LCN13 (By similarity). Acts as a negative regulator of lipogenic gene expression in hepatocytes and thereby mediates the control of very low-density lipoprotein release (PubMed:23307490). May play a role in neurite elongation and survival (By similarity).
TRUSTED_SUSTAINABILITY

Specifications

Antigen TSC22D4
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Tsc22d4
Gene Accession No. Q9Y3Q8
Gene Alias 0610009M14Rik; 1700023B23Rik; AI415410; LOW QUALITY PROTEIN: TSC22 domain family protein 4; THG1; THG-1; Thg1pit; Thg-1pit; Tilz2; TSC22 domain family 4; TSC22 domain family member 4; TSC22 domain family protein 4; TSC22 domain family, member 4; Tsc22d4; tsc-22-like protein THG-1; TSC22-related inducible leucine zipper 2; TSC22-related-inducible leucine zipper protein 2
Gene Symbols Tsc22d4
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human TSC22D4. Recombinant protein control fragment (Product #RP-89392).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 81628
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.