missing translation for 'onlineSavingsMsg'
Learn More

TSC22D4 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

€ 144.20 - € 410.97

Specifications

Antigen TSC22D4
Concentration 0.31 mg/mL
Applications Western Blot, Immunocytochemistry, Immunofluorescence
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16816455
View Documents
Proteintech
55017-1-AP-20UL
20 μL This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
16806455
View Documents
Proteintech
55017-1-AP-150UL
150 μL This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
Description

Description

This antibody is specific to TSC22D4.

ZNF289 (zinc finger protein 289), also known as ARFGAP2 (ADP-ribosylation factor GTPase activating protein 2), IRZ, Zfp289 or Nbla10535, functions as a GTPase-activating protein (GAP) for ARF family proteins. Localizing to the cytoplasmic side of the Golgi apparatus, ZNF289 contains one ARF-GAP domain and is found associated with COP-I-coated vesicles.
Specifications

Specifications

TSC22D4
Western Blot, Immunocytochemistry, Immunofluorescence
Unconjugated
Rabbit
Rat, Human, Mouse
Q3B8N7, Q9EQN3, Q9Y3Q8
684980, 78829, 81628
Peptide
Primary
-20°C
TSC22D4
0.31 mg/mL
Polyclonal
Liquid
RUO
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
THG 1, THG1, TILZ2, Tsc 22 like protein THG 1, TSC22 domain family protein 4, TSC22 domain family, member 4, TSC22D4
Tsc22d4
IgG
Antigen Affinity Chromatography
Antibody
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.