missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ TSEN15 Polyclonal Antibody
GREENER_CHOICE

Product Code. 15754001 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15754001 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15754001 Supplier Invitrogen™ Supplier No. PA556087

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Immunogen sequence: EERGDSEPTP GCSGLGPDGV RGFGDGGGAP SWAPEDAWMG THPKYLEMME LDIGDATHVY VAFLVYLDLM ESK Highest antigen sequence identity to the following orthologs: Mouse - 84%, Rat - 84%.

Non-catalytic subunit of the tRNA-splicing endonuclease complex, a complex responsible for identification and cleavage of the splice sites in pre-tRNA. It cleaves pre-tRNA at the 5' and 3' splice sites to release the intron. The products are an intron and two tRNA half-molecules bearing 2',3' cyclic phosphate and 5'-OH termini. There are no conserved sequences at the splice sites, but the intron is invariably located at the same site in the gene, placing the splice sites an invariant distance from the constant structural features of the tRNA body. The tRNA splicing endonuclease is also involved in mRNA processing via its association with pre-mRNA 3'-end processing factors, establishing a link between pre-tRNA splicing and pre-mRNA 3'-end formation, suggesting that the endonuclease subunits function in multiple RNA-processing events.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TSEN15
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Tsen15
Gene Accession No. Q8WW01
Gene Alias 5730449L18Rik; AL023077; C16H1ORF19; C1orf19; HsSEN15; PCH2F; RGD1308584; sen15; SEN15 homolog; tRNA splicing endonuclease 15 homolog; tRNA splicing endonuclease 15 homolog (S. cerevisiae); tRNA splicing endonuclease subunit 15; tRNA-intron endonuclease Sen15; tRNA-splicing endonuclease subunit Sen15; Tsen15; TSEN15 tRNA splicing endonuclease subunit; Unknown (protein for MGC:127982)
Gene Symbols Tsen15
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human TSEN15. Recombinant protein control fragment (Product #RP-94557).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 116461
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.