missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TSFM Antibody (S06-6H3), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-19981-50ul
This item is not returnable.
View return policy
Description
TSFM Monoclonal antibody specifically detects TSFM in Human samples. It is validated for Western Blot
Specifications
| TSFM | |
| Monoclonal | |
| 0.3 mg/mL | |
| 50mM Tris-Glycine(pH 7.4), 0.15M NaCl, 40% Glycerol, 0.05% BSA | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot | |
| S06-6H3 | |
| Unconjugated | |
| COXPD3, EF-Ts, EFTS, EF-TsMt, EFTSMT, mitochondrial, mitochondrial elongation factor Ts, Ts translation elongation factor, mitochondrial | |
| A synthetic peptide of human TSFM (Uniprot # P43897) | |
| 50 μg | |
| Endocrinology | |
| 10102 | |
| Store at 4°C short term. Store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction