missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TSG Polyclonal Antibody

Rabbit Polyclonal Antibody
€ 184.00 - € 485.00
Specifica
| Antigen | TSG |
|---|---|
| Concentration | 0.5 mg/mL |
| Content And Storage | Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Codice del prodotto | Marca | Quantity | Prezzo | Quantità e disponibilità | |||||
|---|---|---|---|---|---|---|---|---|---|
| Codice del prodotto | Marca | Quantity | Prezzo | Quantità e disponibilità | |||||
17760894
![]() |
Invitrogen™
PA5119492 |
200 μL |
€ 485.00
200µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
17702924
|
Invitrogen™
PA5121522 |
20 μL |
€ 184.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Descrizione
TSG Polyclonal Antibody for Western Blot, IHC (P)
TWSG1 may be involved in dorsoventral axis formation. Seems to antagonize BMP signaling by forming ternary complexes with CHRD and BMPs, thereby preventing BMPs from binding to their receptors. In addition to the anti-BMP function, also has pro-BMP activity, partly mediated by cleavage and degradation of CHRD, which releases BMPs from ternary complexes. May be an important modulator of BMP-regulated cartilage development and chondrocyte differentiation. May play a role in thymocyte development.Specifica
| TSG | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Polyclonal | |
| Liquid | |
| IgG | |
| Human | |
| Twsg1 | |
| 1810013J15Rik; 9030422N06Rik; AW552143; D17Ertd403e; PSEC0250; Tsg; Twg; twisted gastrulation BMP signaling modulator 1; twisted gastrulation homolog 1; twisted gastrulation homolog 1 (Drosophila); twisted gastrulation protein homolog 1; Twsg1 | |
| Twsg1 | |
| Primary | |
| Antigen affinity chromatography, Protein A |
| 0.5 mg/mL | |
| Immunohistochemistry (Paraffin), Western Blot | |
| Unconjugated | |
| Rabbit | |
| RUO | |
| PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4 | |
| Q9GZX9 | |
| 57045 | |
| Recombinant protein Twisted Gastrulation Protein Homolog 1. The antigen corresponds to amino acid range 26-223 of the target protein. | |
| Antibody |
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto