missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TSH beta Antibody (154) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB100-62385
This item is not returnable.
View return policy
Description
TSH beta Monoclonal specifically detects TSH beta in Human samples. It is validated for ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.
Specifications
| TSH beta | |
| Monoclonal | |
| 1.0 mg/mL | |
| ELISA 1:100-1:2000, Immunohistochemistry, Immunocytochemistry/Immunofluorescence | |
| thyroid stimulating hormone, beta, thyroid-stimulating hormone subunit beta, thyrotropin beta chain, thyrotropin beta subunit, thyrotropin subunit beta, TSH-B, TSH-BETA | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| 7252 | |
| Human | |
| Purified |
| ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
| 154 | |
| Unconjugated | |
| P01222 | |
| TSHB | |
| Native Human beta-TSH | |
| 0.5 mg | |
| Primary | |
| This product is specific for is specific for beta thyroid stimulating hormone (beta TSH), a hormone produced by the pituitary gland, which acts to stimulate the growth of the thyroid and production of thyroid hormones. This antibody shows minimal cross-reactivity with beta Human chorionic gonadotropin (hCG), Luteinizing Hormone (LH) or Follicle Stimulating Hormone (FSH). | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction