missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TSH beta Antibody (SR1242), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-21897-50ul
This item is not returnable.
View return policy
Description
TSH beta Monoclonal antibody specifically detects TSH beta in Human samples. It is validated for Western Blot, Immunohistochemistry
Specifications
| TSH beta | |
| Monoclonal | |
| Unconjugated | |
| PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot, Immunohistochemistry | |
| SR1242 | |
| Western Blot 1:500-1:2000, Immunohistochemistry 1:50-1:200 | |
| thyroid stimulating hormone, beta, thyroid-stimulating hormone subunit beta, thyrotropin beta chain, thyrotropin beta subunit, thyrotropin subunit beta, TSH-B, TSH-BETA | |
| A synthesized peptide derived from human TSH beta (Uniprot #: P01222) | |
| 50 μg | |
| Cancer, Signal Transduction | |
| 7252 | |
| Store at -20° C. Avoid freeze/thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction