missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TSH beta Antibody (TSH-116), mFluor Violet 500 SE, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB500-363MFV500
This item is not returnable.
View return policy
Description
TSH beta Monoclonal antibody specifically detects TSH beta in Human samples. It is validated for ELISA, Radioimmune Assays
Specifications
| TSH beta | |
| Monoclonal | |
| mFluor Violet 500 SE | |
| thyroid stimulating hormone, beta, thyroid-stimulating hormone subunit beta, thyrotropin beta chain, thyrotropin beta subunit, thyrotropin subunit beta, TSH-B, TSH-BETA | |
| Human thyrotropin. | |
| 0.1 mL | |
| Cancer, Signal Transduction | |
| 7252 | |
| Store at 4°C in the dark. | |
| IgG2a |
| ELISA | |
| TSH-116 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction