missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TSHR Antibody (484404), Biotin, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB65342B
This item is not returnable.
View return policy
Description
TSHR Monoclonal antibody specifically detects TSHR in Human samples. It is validated for Flow Cytometry
Specifications
| TSHR | |
| Monoclonal | |
| Biotin | |
| CHNG1, LGR3hTSHR-I, MGC75129, seven transmembrane helix receptor, thyroid stimulating hormone receptor, Thyroid-stimulating hormone receptor, thyrotropin receptor, thyrotropin receptor-I, hTSHR-I, TSH-R | |
| NS0 mouse myeloma cell line transfected with human TSH R, Accession # P16473 | |
| 0.1 mL | |
| Cell Cycle and Replication, GPCR, Membrane Trafficking and Chaperones, Neuroscience | |
| 7253 | |
| Store at 4°C in the dark. | |
| IgG1 |
| Flow Cytometry | |
| 484404 | |
| PBS | |
| Mouse | |
| Protein A or G purified from ascites | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction