missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TSHR Antibody (4C1), DyLight 350, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB100-65797UV-0.1ML
This item is not returnable.
View return policy
Description
TSHR Monoclonal specifically detects TSHR in Human samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.
Specifications
| TSHR | |
| Monoclonal | |
| DyLight 350 | |
| 50 mM sodium borate with 0.05% sodium azide | |
| TSHR | |
| External domain of human TSH receptor. | |
| Primary | |
| Recognizes the human thyroid stimulating hormone (TSH) receptor (otherwise known as the thyrotropin receptor) binding to an extracellular domain. | |
| Store at 4°C in the dark. |
| Flow Cytometry, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen) | |
| 4C1 | |
| Flow Cytometry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen | |
| CHNG1, LGR3hTSHR-I, MGC75129, seven transmembrane helix receptor, thyroid stimulating hormone receptor, Thyroid-stimulating hormone receptor, thyrotropin receptor, thyrotropin receptor-I, hTSHR-I, TSH-R | |
| Mouse | |
| 0.1 mL | |
| 7253 | |
| Human | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction