Learn More
TSHR Antibody (4C1), DyLight 680, Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NB100-65797FR-0.1ML
Product Code. 18074554
  • ONLINE EXCLUSIVE PRICE
    € 659.00
    € 622.65 / 0.10mL
    Save € 36.35
    5% Off
Description
TSHR Monoclonal specifically detects TSHR in Human samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
TSHR
Monoclonal
DyLight 680
50 mM sodium borate with 0.05% sodium azide
TSHR
External domain of human TSH receptor.
Primary
Recognizes the human thyroid stimulating hormone (TSH) receptor (otherwise known as the thyrotropin receptor) binding to an extracellular domain.
Store at 4°C in the dark.
Flow Cytometry, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen)
4C1
Flow Cytometry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen
CHNG1, LGR3hTSHR-I, MGC75129, seven transmembrane helix receptor, thyroid stimulating hormone receptor, Thyroid-stimulating hormone receptor, thyrotropin receptor, thyrotropin receptor-I, hTSHR-I, TSH-R
Mouse
0.1 mL
7253
Human
IgG2a