Learn More
TSHZ1 Monoclonal Antibody (OTI2G9), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA809958
Description
The Teashirt zinc finger homeobox (TSHZ) family comprise a family of evolutionarily conserved transcription factors that, in Drosophila, are active in specific body parts for patterning, but whose function in vertebrates is less clear. TSHZ1 has been found to be required for axial skeleton, soft palate and middle ear development in mice and may be involved in a common pathway with the Hox genes. Both TSHZ1 and the related protein TSHZ3 have been found to interact with FE65, an adapter protein that binds to the amyloid protein precursor (APP) in neurons. Together with SET, a component of the inhibitor of acetyl transferase, and histone deacetylases, these proteins formed a gene-silencing complex whose target includes caspase-4.Specifications
| TSHZ1 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q6ZSZ6 | |
| TSHZ1 | |
| Human recombit protein fragment corresponding to amino acids 29-176 of human TSHZ1 produced in E.coli. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Western Blot | |
| OTI2G9 | |
| Unconjugated | |
| TSHZ1 | |
| CAA, NY-CO-33, SDCCAG33, TSH1 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 10194 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Le programme Encompass de Fisher Scientific propose des articles qui ne font pas partie de notre portefeuille de distribution. Ces produits n'ont généralement pas de photos ou de descriptions détaillées. Cependant, nous nous engageons à améliorer votre expérience d'achat. Veuillez utiliser le formulaire ci-dessous pour nous faire part de vos commentaires sur le contenu de ce produit.