missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TSHZ1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA534342
This item is not returnable.
View return policy
Description
A suggested positive control is A-20 cell lysate. PA5-34342 can be used with blocking peptide PEP-1385.
This gene encodes a colon cancer antigen that was defined by serological analysis of recombinant cDNA expression libraries. The encoded protein is a member of the teashirt C2H2-type zinc-finger protein family and may be involved in transcriptional regulation of developmental processes.
Specifications
| TSHZ1 | |
| Polyclonal | |
| Unconjugated | |
| TSHZ1 | |
| 5730407I04Rik; antigen NY-CO-33; CAA; D18Bwg1409e; Kiaa4206; mKIAA4206; Mtsh1; NY-CO-33; SDCCAG33; Serologically defined colon cancer antigen 3 homolog; serologically defined colon cancer antigen 33; Teashirt homolog 1; teashirt zinc finger family member 1; teashirt zinc finger homeobox 1; teashirt1; TSH1; Tshz1 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 10194, 110796, 307217 | |
| Maintain refrigerated at 2°C to 8°C for up to 3 months. For long term storage store at -20°C | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 0.02% sodium azide | |
| Q5DTH5, Q6ZSZ6 | |
| TSHZ1 | |
| A 16 amino acid synthetic peptide near the amino terminus of human TSHZ1. | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction