missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TSLP Antibody (55N1D10), Janelia Fluor™ 549, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-27326JF549
This item is not returnable.
View return policy
Description
TSLP Monoclonal antibody specifically detects TSLP in Human, Rabbit samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
| TSLP | |
| Monoclonal | |
| Janelia Fluor 549 | |
| thymic stromal lymphopoietin | |
| A partial recombinant TSLP protein (amino acids 27-158) was used as the immunogen for the antibody. | |
| 0.1 mL | |
| Biologically Active Proteins, Innate Immunity | |
| 85480 | |
| Store at 4°C in the dark. | |
| IgG2a κ |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| 55N1D10 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| Human, Rabbit | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction