missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TSLP Antibody (55N1E3) [CoraFluor™ 1], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP2-27323CL1
This item is not returnable.
View return policy
Description
TSLP Monoclonal antibody specifically detects TSLP in Human samples. It is validated for Western Blot
Specifications
| TSLP | |
| Monoclonal | |
| CoraFluor 1 | |
| thymic stromal lymphopoietin | |
| amino acids (27-158) MDYDFTNCDFEkIkAAYLSTISkDLITYMSGTkSTEFNNTVSCSNRPHCLTEIQSLTFNP∽TAGCASLAkEMFAMkTkAALAIWCPGYSETQINATQAMkkRRkRkVTTNkCLEQVSQLQG∽LWRRFNRPLLkQQL | |
| 0.1 mL | |
| Biologically Active Proteins, Innate Immunity | |
| 85480 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG1 κ |
| Western Blot | |
| 55N1E3 | |
| PBS | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction