missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TSLP Antibody, DyLight 488, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB110-55233G
This item is not returnable.
View return policy
Description
TSLP Polyclonal antibody specifically detects TSLP in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
| TSLP | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Rabbit | |
| Peptide affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| DyLight 488 | |
| thymic stromal lymphopoietin | |
| Antibody was raised against a peptide corresponding to 17 amino acids near the carboxy terminus of human TSLP. The immunogen is located within the last 50 amino acids of TSLP. Amino Acid Sequence: ATQAMKKRRKRKVTTNK | |
| 0.1 mL | |
| Biologically Active Proteins, Innate Immunity | |
| 85480 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction