missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ THRA Recombinant Rabbit Monoclonal Antibody (0O4G3)
GREENER_CHOICE

Product Code. 30283050
Change view
Click to view available options
Quantity:
20 μL
100 μL
Unit Size:
100µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30283050 100 μL 100µL
30282453 20 μL 20µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30283050 Supplier Invitrogen™ Supplier No. MA557902

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Recombinant Monoclonal Antibody

Immunogen sequence: GMAMDLVLDD SKRVAKRKLI EQNRERRRKE EMIRSLQQRP EPTPEEWDLI HIATEAHRST NAQGSHWKQR RKFLPDDIGQ S.

The protein encoded by this gene is a nuclear hormone receptor for triiodothyronine. It is one of the several receptors for thyroid hormone, and has been shown to mediate the biological activities of thyroid hormone. Knockout studies in mice suggest that the different receptors, while having certain extent of redundancy, may mediate different functions of thyroid hormone. Alternatively spliced transcript variants encoding distinct isoforms have been reported.
TRUSTED_SUSTAINABILITY

Specifications

Antigen THRA
Applications ELISA, Western Blot
Classification Recombinant Monoclonal
Clone 0O4G3
Concentration 1.13 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.05% BSA, 50% glycerol and 0.05% ProClin 300; pH 7.3
Gene Thra
Gene Accession No. P10827, P63058
Gene Alias 6430529J03Rik; AR7; avian erythroblastic leukemia viral (v-erb-a) oncogene homolog 1; AW259572; cerbA protein; c-erb-A protein; c-erb-A thyroid hormone receptor; c-ERBA-1; cerbAalpha; c-erbAalpha; c-erbA-alpha; c-erbAalpha2; c-erbA-alpha2; c-erbAalpha2; thyroid hormone receptor alpha; CHNG6; E ERB-T-1; EAR7; EAR-7; ERBA; ERBA ALPHA; ERBA related 7; ERBA1; ERBA-related 7; ERB-T-1; MGC000261; MGC43240; NR1A1; Nuclear receptor subfamily 1 group A member 1; Oncogene ERBA; Rvr; T3R[a]; T3Ralpha; Thra; THRA1; THRA2; Thyroid hormone receptor alpha; thyroid hormone receptor alpha 1; thyroid hormone receptor alpha 1 (avian erythroblastic leukemia viral (v-erb-a) oncogene homolog 1 formerly ERBA1); thyroid hormone receptor alpha 2; thyroid hormone receptor beta; thyroid hormone receptor, alpha; thyroid hormone receptor, alpha (erythroblastic leukemia viral (v-erb-a) oncogene homolog, avian); thyroid normone nuclear receptor alpha variant 1; TR; TR alpha 1; TR alpha 1+2; TR alpha 2; triiodothyronine receptor; unnamed protein product; V-erbA-related protein 7
Gene Symbols Thra
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence within amino acids 119-199 of human THRA (NP_003241.2).
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 21833, 7067
Target Species Human, Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.