missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
May be involved in proteolysis through its threonine endopeptidase activity.
Specifications
Specifications
| Antigen | TSP50 |
| Applications | Immunohistochemistry (Paraffin), Immunofluorescence, Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 0.33 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 |
| Gene | PRSS50 |
| Gene Accession No. | Q8BLH5, Q9UI38 |
| Gene Alias | Cancer/testis antigen 20, CT20, PRSS50, Serine protease 50, testes protease 50, TSP50 |
| Gene Symbols | PRSS50 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?