missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TSPAN1 Antibody (819202) [Janelia Fluor™ 549], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB9186I
This item is not returnable.
View return policy
Description
TSPAN1 Monoclonal antibody specifically detects TSPAN1 in Human samples. It is validated for Flow Cytometry
Specifications
| TSPAN1 | |
| Monoclonal | |
| Janelia Fluor 549 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 10103 | |
| Store at 4°C in the dark. | |
| IgG3 |
| Flow Cytometry | |
| 819202 | |
| Flow Cytometry | |
| NET1, NET-1, tetraspan 1, Tetraspan NET-1, tetraspanin 1, Tetraspanin TM4-C, tetraspanin-1, TM4C, TM4SF, TSPAN-1 | |
| Mouse myeloma cell line NS0-derived recombinant human TSPAN1, Met1-Gln241, Accession # O60635 | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction