missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TSPAN10 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA543482
This item is not returnable.
View return policy
Description
Peptide sequence: SCVKYLIFLS NFPFSLLGLL ALAIGLWGLA VKGSLGSDLG GPLPADPMLG Sequence homology: Human: 100%.
TSPAN10 belongs to the tetraspanin (TM4SF) family. It is a multi-pass membrane protein. The exact function of TSPAN10 remains unknown.
Specifications
| TSPAN10 | |
| Polyclonal | |
| Unconjugated | |
| TSPAN10 | |
| OCSP; oculospanin; tetraspanin 10; tetraspanin-10; Tspan10; Tspan-10 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 83882 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q9H1Z9 | |
| TSPAN10 | |
| Synthetic peptide directed towards the N-terminal of human TSPAN10. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction