missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TSPAN13 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA533092
This item is not returnable.
View return policy
Description
Percent identity with other species by BLAST analysis: Human, Gorilla, Gibbon, Monkey, Marmoset, Mouse, Rat, Dog, Elephant, Panda, Horse, Rabbit (100%).
The protein encoded by this gene is a member of the transmembrane 4 superfamily, also known as the tetraspanin family. Most of these members are cell-surface proteins that are characterized by the presence of four hydrophobic domains. The proteins mediate signal transduction events that play a role in the regulation of cell development, activation, growth and motility.
Specifications
| TSPAN13 | |
| Polyclonal | |
| Unconjugated | |
| TSPAN13 | |
| 1100001I23Rik; NET6; NET-6; tetraspan NET-6; Tetraspan Tspan-13; tetraspanin 13; tetraspanin-13; Tm4sf13; transmembrane 4 superfamily member 13; transmembrane 4 superfamily member tetraspan NET-6; Tspan13; tspan-13; UNQ260/PRO296 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 100066103, 100358391, 27075, 366602, 482332, 66109 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.9 mg/mL | |
| PBS with 0.1% sodium azide | |
| O95857, Q5FVL6, Q9D8C2 | |
| TSPAN13 | |
| Synthetic 13 amino acid peptide from internal region of human TSPAN13. | |
| 50 μg | |
| Primary | |
| Human, Mouse, Rat, Canine, Rabbit, Monkey, Horse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction