missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TSPAN2 Antibody (822509), PE/Cy5.5, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB7876PECY55
This item is not returnable.
View return policy
Description
TSPAN2 Monoclonal antibody specifically detects TSPAN2 in Human samples. It is validated for Flow Cytometry
Specifications
| TSPAN2 | |
| Monoclonal | |
| PE-Cyanine5.5 | |
| PBS | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 10100 | |
| Store at 4C in the dark. Do not freeze. | |
| IgG2b |
| Flow Cytometry | |
| 822509 | |
| Flow Cytometry | |
| FLJ12082, NET3, new EST tetraspan 3, tetraspan 2, Tetraspan NET-3, tetraspan TM4SF, tetraspanin 2, tetraspanin 2 isoform, tetraspanin-2, TSN2, TSPAN-2 | |
| NS0 mouse myeloma cell line transfected with human TSPAN2, Met1-Ile221, Accession # O60636 | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction